missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7129904
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
F07955G 5 g
F079525G 25 g
2 options

Catalog No. F07955G

Supplier: TCI America F07955G

Only null left

Chemical Identifiers

CAS 2905-21-7
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042285
InChI Key ZSZKAQCISWFDCQ-UHFFFAOYSA-N
Synonym 2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
PubChem CID 137761
IUPAC Name 2-fluorobenzene-1-sulfonyl chloride
SMILES FC1=CC=CC=C1S(Cl)(=O)=O

Specifications

Boiling Point 127°C
Quantity 5 g
UN Number 3261
Formula Weight 194.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluorobenzenesulfonyl Chloride