missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzenesulfonyl chloride, 98%

Catalog No. AAA1418106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418106 5 g
AAA1418114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1418106 Supplier Thermo Scientific Chemicals Supplier No. A1418106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2905-21-7 | C6H4ClFO2S | 194.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-21-7
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042285
InChI Key ZSZKAQCISWFDCQ-UHFFFAOYSA-N
Synonym 2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
PubChem CID 137761
IUPAC Name 2-fluorobenzenesulfonyl chloride
SMILES FC1=CC=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 27°C to 32°C
Density 1.47
Boiling Point 246°C to 247°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 5 g
UN Number UN3261
Beilstein 2093979
Sensitivity Moisture sensitive
Formula Weight 194.61
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only