missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 2905-21-7
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042285
InChI Key ZSZKAQCISWFDCQ-UHFFFAOYSA-N
Synonym 2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
PubChem CID 137761
IUPAC Name 2-fluorobenzenesulfonyl chloride
SMILES FC1=CC=CC=C1S(Cl)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC375780010
SDS View Documents
Thermo Scientific Chemicals
375780010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC375780050
SDS View Documents
Thermo Scientific Chemicals
375780050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2905-21-7
194.60
ZSZKAQCISWFDCQ-UHFFFAOYSA-N
137761
FC1=CC=CC=C1S(Cl)(=O)=O
C6H4ClFO2S
MFCD00042285
2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
2-fluorobenzenesulfonyl chloride

Specifications

2905-21-7
100.0
>110°C
96% min. (ex Cl) (Argentometry)
C6H4ClFO2S
FC6H4SO2Cl
1g
ZSZKAQCISWFDCQ-UHFFFAOYSA-N
2-fluorobenzenesulfonyl chloride
137761
97%
96.0
140.0°C to 142.0°C (12.0mmHg)
Authentic
Glass bottle
1.5350 to 1.5380
MFCD00042285
2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
FC1=CC=CC=C1S(Cl)(=O)=O
194.60
194.61
2-Fluorobenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only