missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-N-methylaniline 98.0+%, TCI America™

Catalog No. F044925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F044925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F044925G Supplier TCI America Supplier No. F044925G

Chemical Identifiers

CAS 1978-38-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00028030
InChI Key LDVAIJZDACHGML-UHFFFAOYSA-N
PubChem CID 2759010
IUPAC Name 2-fluoro-N-methylaniline
SMILES CNC1=CC=CC=C1F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-N-methylaniline