missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-DL-phenylalanine, 98%

Catalog No. p-7044666
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0765403 1 g
AAL0765406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0765403 Supplier Thermo Scientific Chemicals Supplier No. L0765403
Only null left

CAS: 2629-55-2 | C9H10FNO2 | 183.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2629-55-2
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00004271
InChI Key NYCRCTMDYITATC-UHFFFAOYSA-N
Synonym 2-fluoro-dl-phenylalanine, 2-amino-3-2-fluorophenyl propanoic acid, 2-fluorophenylalanine, o-fluoro-dl-phenylalanine, o-fluorophenylalanine, 3-o-fluorophenyl alanine, dl-2-fluorophenyl alanine, dl-2-f-phe-oh, dl-o-fluorophenylalanine, dl-beta-o-fluorophenylalanine
PubChem CID 9465
IUPAC Name 2-amino-3-(2-fluorophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CC(C(=O)O)N)F

Specifications

Melting Point ∽245°C (decomposition)
Beilstein 3201186
Quantity 1 g
Formula Weight 183.18
Percent Purity 98%
Chemical Name or Material 2-Fluoro-DL-phenylalanine
EINECSNumber 220-105-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only