missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-9H-carbazole 98.0+%, TCI America™
$162.17 - $484.65
Chemical Identifiers
| CAS | 391-53-7 |
|---|---|
| Molecular Formula | C12H8FN |
| Molecular Weight (g/mol) | 185.20 |
| MDL Number | MFCD09033508 |
| InChI Key | RQGSXFCTBTUNRR-UHFFFAOYSA-N |
| PubChem CID | 11816279 |
| IUPAC Name | 2-fluoro-9H-carbazole |
| SMILES | FC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F1182200MG
|
TCI America
F1182200MG |
200 mg |
Each for $162.17
|
|
|||||
|
F11821G
|
TCI America
F11821G |
1 g |
Each for $484.65
|
|
|||||
Chemical Identifiers
| 391-53-7 | |
| 185.20 | |
| RQGSXFCTBTUNRR-UHFFFAOYSA-N | |
| 2-fluoro-9H-carbazole |
| C12H8FN | |
| MFCD09033508 | |
| 11816279 | |
| FC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 |
Specifications
| 391-53-7 | |
| C12H8FN | |
| 200 mg | |
| FC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 | |
| 185.20 | |
| 185.20 | |
| Crystalline Powder |
| 227°C | |
| MFCD09033508 | |
| RQGSXFCTBTUNRR-UHFFFAOYSA-N | |
| 2-fluoro-9H-carbazole | |
| 11816279 | |
| ≥98.0% (GC) | |
| 2-Fluoro-9H-carbazole |
Safety and Handling
TSCA : No