missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-methylpyridine 95.0+%, TCI America™
$32.72 - $88.56
Chemical Identifiers
| CAS | 407-22-7 |
|---|---|
| Molecular Formula | C6H6FN |
| Molecular Weight (g/mol) | 111.12 |
| MDL Number | MFCD00041226 |
| InChI Key | UDMNVTJFUISBFD-UHFFFAOYSA-N |
| Synonym | 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga |
| PubChem CID | 96090 |
| IUPAC Name | 2-fluoro-6-methylpyridine |
| SMILES | CC1=CC=CC(F)=N1 |
Chemical Identifiers
| 407-22-7 | |
| 111.12 | |
| UDMNVTJFUISBFD-UHFFFAOYSA-N | |
| 96090 | |
| CC1=CC=CC(F)=N1 |
| C6H6FN | |
| MFCD00041226 | |
| 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga | |
| 2-fluoro-6-methylpyridine |
Specifications
| 407-22-7 | |
| 142°C | |
| MFCD00041226 | |
| 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga | |
| CC1=CC=CC(F)=N1 | |
| 111.12 | |
| 111.12 | |
| Liquid |
| Yellow | |
| C6H6FN | |
| 1 g | |
| UDMNVTJFUISBFD-UHFFFAOYSA-N | |
| 2-fluoro-6-methylpyridine | |
| 96090 | |
| ≥95.0% (GC) | |
| 2-Fluoro-6-methylpyridine |
Safety and Handling
TSCA : No