missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylpyridine 95.0+%, TCI America™

Catalog No. F05201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05201G 1 g
F05205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05201G Supplier TCI America Supplier No. F05201G

Chemical Identifiers

CAS 407-22-7
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00041226
InChI Key UDMNVTJFUISBFD-UHFFFAOYSA-N
Synonym 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga
PubChem CID 96090
IUPAC Name 2-fluoro-6-methylpyridine
SMILES CC1=CC=CC(F)=N1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 1 g
Formula Weight 111.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-methylpyridine