missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylpyridine, 98%

CAS: 407-22-7 | C6H6FN | 111.12 g/mol

$34.59 - $115.99

Chemical Identifiers

CAS 407-22-7
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00041226
InChI Key UDMNVTJFUISBFD-UHFFFAOYSA-N
Synonym 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga
PubChem CID 96090
IUPAC Name 2-fluoro-6-methylpyridine
SMILES CC1=CC=CC(F)=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0156103
View Documents
Thermo Scientific Chemicals
L0156103
1 g
Each for $34.59
Only null left
 
AAL0156106
View Documents
Thermo Scientific Chemicals
L0156106
5 g
Each for $115.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

407-22-7
111.12
UDMNVTJFUISBFD-UHFFFAOYSA-N
96090
CC1=CC=CC(F)=N1
C6H6FN
MFCD00041226
2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga
2-fluoro-6-methylpyridine

Specifications

407-22-7
140°C to 141°C
C6H6FN
MFCD00041226
107084
UDMNVTJFUISBFD-UHFFFAOYSA-N
2-fluoro-6-methylpyridine
96090
98%
1.077
66°C (150°F)
1.47
1 g
2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga
CC1=CC=CC(F)=N1
111.12
111.12
2-Fluoro-6-methylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 206-980-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only