missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylpyridine, 98%

Catalog No. AAL0156103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0156103 1 g
AAL0156106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0156103 Supplier Thermo Scientific Chemicals Supplier No. L0156103

CAS: 407-22-7 | C6H6FN | 111.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 407-22-7
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00041226
InChI Key UDMNVTJFUISBFD-UHFFFAOYSA-N
Synonym 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga
PubChem CID 96090
IUPAC Name 2-fluoro-6-methylpyridine
SMILES CC1=CC=CC(F)=N1

Specifications

Density 1.077
Boiling Point 140°C to 141°C
Flash Point 66°C (150°F)
Refractive Index 1.47
Quantity 1 g
Beilstein 107084
Formula Weight 111.12
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-980-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only