missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$50.33 - $50.33
Chemical Identifiers
| CAS | 78495-63-3 |
|---|---|
| Molecular Formula | C7H8BFO3 |
| MDL Number | MFCD02179483 |
| InChI Key | XOVMDVZAWWQSDC-UHFFFAOYSA-N |
| PubChem CID | 3294524 |
| IUPAC Name | (2-fluoro-6-methoxyphenyl)boronic acid |
Chemical Identifiers
| 78495-63-3 | |
| MFCD02179483 | |
| 3294524 |
| C7H8BFO3 | |
| XOVMDVZAWWQSDC-UHFFFAOYSA-N | |
| (2-fluoro-6-methoxyphenyl)boronic acid |
Specifications
| 78495-63-3 | |
| White | |
| MFCD02179483 | |
| 2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef | |
| B(C1=C(C=CC=C1F)OC)(O)O | |
| 169.946 | |
| 169.95 | |
| 2-Fluoro-6-methoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 125°C | |
| C7H8BFO3 | |
| 1 g | |
| XOVMDVZAWWQSDC-UHFFFAOYSA-N | |
| (2-fluoro-6-methoxyphenyl)boronic acid | |
| 3294524 | |
| Crystalline Powder |
Safety and Handling
TSCA : No