missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F09411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09411G Supplier TCI America Supplier No. F09411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78495-63-3
Molecular Formula C7H8BFO3
MDL Number MFCD02179483
InChI Key XOVMDVZAWWQSDC-UHFFFAOYSA-N
PubChem CID 3294524
IUPAC Name (2-fluoro-6-methoxyphenyl)boronic acid

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Synonym 2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef
SMILES B(C1=C(C=CC=C1F)OC)(O)O
Molecular Weight (g/mol) 169.946
Formula Weight 169.95
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-methoxyphenylboronic Acid (contains varying amounts of Anhydride)