missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzeneboronic acid, 98%

CAS: 78495-63-3 | C7H8BFO3 | 169.946 g/mol

$30.53 - $102.55

Chemical Identifiers

CAS 78495-63-3
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD02179483
InChI Key XOVMDVZAWWQSDC-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef
PubChem CID 3294524
IUPAC Name (2-fluoro-6-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC=C1F)OC)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2606403
View Documents
Thermo Scientific Chemicals
H2606403
1 g
Each for $30.53
Only null left
 
AAH2606406
View Documents
Thermo Scientific Chemicals
H2606406
5 g
Each for $102.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78495-63-3
169.946
XOVMDVZAWWQSDC-UHFFFAOYSA-N
3294524
B(C1=C(C=CC=C1F)OC)(O)O
C7H8BFO3
MFCD02179483
2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef
(2-fluoro-6-methoxyphenyl)boronic acid

Specifications

78495-63-3
C7H8BFO3
1 g
XOVMDVZAWWQSDC-UHFFFAOYSA-N
(2-fluoro-6-methoxyphenyl)boronic acid
3294524
98%
124°C to 126°C
MFCD02179483
2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef
B(C1=C(C=CC=C1F)OC)(O)O
169.946
169.95
2-Fluoro-6-methoxybenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only