missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzeneboronic acid, 98%

Catalog No. AAH2606403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2606403 1 g
AAH2606406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2606403 Supplier Thermo Scientific Chemicals Supplier No. H2606403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78495-63-3 | C7H8BFO3 | 169.946 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78495-63-3
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD02179483
InChI Key XOVMDVZAWWQSDC-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef
PubChem CID 3294524
IUPAC Name (2-fluoro-6-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC=C1F)OC)(O)O

Specifications

Melting Point 124°C to 126°C
Quantity 1 g
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only