missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-hydroxypyridine 98.0+%, TCI America™

Catalog No. F09801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09801G 1 g
F09805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09801G Supplier TCI America Supplier No. F09801G

Chemical Identifiers

CAS 55758-32-2
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD01075040
InChI Key HTRLNWYWOKWCLV-UHFFFAOYSA-N
Synonym 2-fluoro-5-hydroxypyridine, 6-fluoro-3-pyridinol, 6-fluoro-3-hydroxypyridine, 3-pyridinol, 6-fluoro, pubchem6321, 6-fluoropyridine-3-ol, 6-fluoro-pyridin-3-ol, acmc-1b21k, 3-hydroxy-6-fluoro-pyridine, ksc490s4n
PubChem CID 2763108
IUPAC Name 6-fluoropyridin-3-ol
SMILES C1=CC(=NC=C1O)F

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Formula Weight 113.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-hydroxypyridine