missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. F09951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09951G 1 g
F09955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09951G Supplier TCI America Supplier No. F09951G

Chemical Identifiers

CAS 118078-66-3
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD09879262
InChI Key DFNQBXZKPUBEIX-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl pyridine, 2-fluoro-4-trifluoromethyl-pyridine, pyridine, 2-fluoro-4-trifluoromethyl, pubchem23563, acmc-1c7kk, 2-fluoranyl-4-trifluoromethyl pyridine
PubChem CID 11159514
IUPAC Name 2-fluoro-4-(trifluoromethyl)pyridine
SMILES C1=CN=C(C=C1C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
UN Number 1992
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)pyridine