Learn More
2-Fluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
$192.42 - $192.42
Chemical Identifiers
| CAS | 118078-66-3 |
|---|---|
| Molecular Formula | C6H3F4N |
| Molecular Weight (g/mol) | 165.091 |
| MDL Number | MFCD09879262 |
| InChI Key | DFNQBXZKPUBEIX-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-trifluoromethyl pyridine, 2-fluoro-4-trifluoromethyl-pyridine, pyridine, 2-fluoro-4-trifluoromethyl, pubchem23563, acmc-1c7kk, 2-fluoranyl-4-trifluoromethyl pyridine |
| PubChem CID | 11159514 |
| IUPAC Name | 2-fluoro-4-(trifluoromethyl)pyridine |
| SMILES | C1=CN=C(C=C1C(F)(F)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3319806
|
Thermo Scientific Chemicals
H3319806 |
5 g |
Each for $192.42
|
|
|||||
Description
2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.
Solubility
Miscible with water, alcohol, acetonitrile and N,N-dimethylformamide
Notes
Incompatible with oxidizing agents and bases.
Chemical Identifiers
| 118078-66-3 | |
| 165.091 | |
| DFNQBXZKPUBEIX-UHFFFAOYSA-N | |
| 11159514 | |
| C1=CN=C(C=C1C(F)(F)F)F |
| C6H3F4N | |
| MFCD09879262 | |
| 2-fluoro-4-trifluoromethyl pyridine, 2-fluoro-4-trifluoromethyl-pyridine, pyridine, 2-fluoro-4-trifluoromethyl, pubchem23563, acmc-1c7kk, 2-fluoranyl-4-trifluoromethyl pyridine | |
| 2-fluoro-4-(trifluoromethyl)pyridine |
Specifications
| 118078-66-3 | |
| C6H3F4N | |
| 5 g | |
| Miscible with water,alcohol,acetonitrile and N,N-dimethylformamide | |
| C1=CN=C(C=C1C(F)(F)F)F | |
| 165.091 | |
| 165.09 | |
| 2-Fluoro-4-(trifluoromethyl)pyridine |
| 113°C | |
| MFCD09879262 | |
| 2-fluoro-4-trifluoromethyl pyridine, 2-fluoro-4-trifluoromethyl-pyridine, pyridine, 2-fluoro-4-trifluoromethyl, pubchem23563, acmc-1c7kk, 2-fluoranyl-4-trifluoromethyl pyridine | |
| DFNQBXZKPUBEIX-UHFFFAOYSA-N | |
| 2-fluoro-4-(trifluoromethyl)pyridine | |
| 11159514 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only