missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%

Catalog No. AAH3320303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3320303 1 g
AAH3320306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3320303 Supplier Thermo Scientific Chemicals Supplier No. H3320303

CAS: 1227628-83-2 | C8H4F4O2 | 208.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1227628-83-2
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD16652440
InChI Key DEACWPDDLIOZJV-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethoxy benzaldehyde
PubChem CID 53465593
IUPAC Name 2-fluoro-4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)F)C=O

Specifications

Refractive Index 1.4455
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only