missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. p-7054484
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62718MD 250 mg
AAH6271803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62718MD Supplier Thermo Scientific Chemicals Supplier No. H62718MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073353-89-5
Molecular Formula C12H15BFNO4
Molecular Weight (g/mol) 267.063
MDL Number MFCD09264075
InChI Key QYXHQOSFZITWSU-UHFFFAOYSA-N
Synonym 2-2-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-nitrophenylboronic acid, pinacol ester, 2-fluoro-4-nitrophenylboronic acid,pinacol ester
PubChem CID 46738935
IUPAC Name 2-(2-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

Quantity 250 mg
Formula Weight 267.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-nitrobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only