missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylpyridine, 98%

Catalog No. AAL0160203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0160203 1 g
AAL0160206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0160203 Supplier Thermo Scientific Chemicals Supplier No. L0160203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 461-87-0 | C6H6FN | 111.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-87-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041224
InChI Key ZBFAXMKJADVOGH-UHFFFAOYSA-N
Synonym 2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j
PubChem CID 2737403
IUPAC Name 2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1)F

Specifications

Density 1.078
Boiling Point 155°C to 156°C
Flash Point 51°C (123°F)
Refractive Index 1.4725
Quantity 1 g
UN Number UN1993
Beilstein 107086
Formula Weight 111.12
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only