missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxyphenylboronic acid, 97%, Thermo Scientific™

Catalog No. p-4812798
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449840010 1g Glass bottle
AC449840050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449840010 Supplier Thermo Scientific Chemicals Supplier No. 449840010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 162101-31-7
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
InChI Key ULUIXJDBPYBAHS-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 4-borono-3-fluoroanisole, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, 2-fluoro-4-methoxyphenylboronicacid, pubchem15367, acmc-209dnv, ksc498a9p, 2-fluoro-4methoxyphenylboronic acid
PubChem CID 15789384
IUPAC Name (2-fluoro-4-methoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OC)F)(O)O

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1g
Formula Weight 169.95
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methoxyphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only