missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxybenzeneboronic acid, 98%

Catalog No. AAL1996003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996003 1 g
AAL1996006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1996003 Supplier Thermo Scientific Chemicals Supplier No. L1996003

CAS: 162101-31-7 | C7H8BFO3 | 169.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162101-31-7
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD04039323
InChI Key ULUIXJDBPYBAHS-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 4-borono-3-fluoroanisole, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, 2-fluoro-4-methoxyphenylboronicacid, pubchem15367, acmc-209dnv, ksc498a9p, 2-fluoro-4methoxyphenylboronic acid
PubChem CID 15789384
IUPAC Name (2-fluoro-4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(F)=C1

Specifications

Quantity 1 g
Beilstein 9192698
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only