missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 263540250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC263540250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

74457-86-6
168.17
PIRRWUMTIBFCCW-UHFFFAOYSA-N
592821
COC1=CC=C(C(C)=O)C(F)=C1
C9H9FO2
MFCD00042290
2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
1-(2-fluoro-4-methoxyphenyl)ethanone

Specifications

2'-Fluoro-4'-methoxyacetophenone
74457-86-6
100.0
>110°C
98.5% min. (GC)
C9H9FO2
25g
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
COC1=CC=C(C(C)=O)C(F)=C1
168.17
168.17
99%
98.5
80.0°C to 85.0°C (0.1mmHg)
Authentic
Glass bottle
MFCD00042290
2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
PIRRWUMTIBFCCW-UHFFFAOYSA-N
1-(2-fluoro-4-methoxyphenyl)ethanone
592821
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...