missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-methoxyacetophenone, 99%

Catalog No. AAA1689106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1689106 5 g
AAA1689114 25 g
AAA1689122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1689106 Supplier Thermo Scientific Chemicals Supplier No. A1689106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74457-86-6 | C9H9FO2 | 168.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74457-86-6
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00042290
InChI Key PIRRWUMTIBFCCW-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
PubChem CID 592821
IUPAC Name 1-(2-fluoro-4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C(C)=O)C(F)=C1

Specifications

Melting Point 50°C to 53°C
Boiling Point 80°C to 85°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 3539992
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material 2'-Fluoro-4'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only