missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-iodopyridine 98.0+%, TCI America™
$72.70 - $279.60
Chemical Identifiers
| CAS | 22282-70-8 |
|---|---|
| Molecular Formula | C5H3FIN |
| Molecular Weight (g/mol) | 222.989 |
| MDL Number | MFCD03092926 |
| InChI Key | ADPRIAVYIGHFSO-UHFFFAOYSA-N |
| Synonym | 4-iodo-2-fluoropyridine, 2-fluoro-4-iodo-pyridine, 2-fluoro-4-iodo pyridine, pyridine, 2-fluoro-4-iodo, pubchem1231, pubchem3510, fluoro-4-iodo pyridine, acmc-1cg64, ksc201s6h |
| PubChem CID | 7023570 |
| IUPAC Name | 2-fluoro-4-iodopyridine |
| SMILES | C1=CN=C(C=C1I)F |
Chemical Identifiers
| 22282-70-8 | |
| 222.989 | |
| ADPRIAVYIGHFSO-UHFFFAOYSA-N | |
| 7023570 | |
| C1=CN=C(C=C1I)F |
| C5H3FIN | |
| MFCD03092926 | |
| 4-iodo-2-fluoropyridine, 2-fluoro-4-iodo-pyridine, 2-fluoro-4-iodo pyridine, pyridine, 2-fluoro-4-iodo, pubchem1231, pubchem3510, fluoro-4-iodo pyridine, acmc-1cg64, ksc201s6h | |
| 2-fluoro-4-iodopyridine |
Specifications
| 22282-70-8 | |
| White-Yellow | |
| MFCD03092926 | |
| 4-iodo-2-fluoropyridine, 2-fluoro-4-iodo-pyridine, 2-fluoro-4-iodo pyridine, pyridine, 2-fluoro-4-iodo, pubchem1231, pubchem3510, fluoro-4-iodo pyridine, acmc-1cg64, ksc201s6h | |
| C1=CN=C(C=C1I)F | |
| 222.989 | |
| 222.99 | |
| Crystalline Powder |
| 58°C | |
| C5H3FIN | |
| 1 g | |
| ADPRIAVYIGHFSO-UHFFFAOYSA-N | |
| 2-fluoro-4-iodopyridine | |
| 7023570 | |
| ≥98.0% (GC) | |
| 2-Fluoro-4-iodopyridine |
Safety and Handling
TSCA : No