missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodopyridine 98.0+%, TCI America™

Catalog No. F07731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07731G 1 g
F07735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07731G Supplier TCI America Supplier No. F07731G

Chemical Identifiers

CAS 22282-70-8
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03092926
InChI Key ADPRIAVYIGHFSO-UHFFFAOYSA-N
Synonym 4-iodo-2-fluoropyridine, 2-fluoro-4-iodo-pyridine, 2-fluoro-4-iodo pyridine, pyridine, 2-fluoro-4-iodo, pubchem1231, pubchem3510, fluoro-4-iodo pyridine, acmc-1cg64, ksc201s6h
PubChem CID 7023570
IUPAC Name 2-fluoro-4-iodopyridine
SMILES C1=CN=C(C=C1I)F

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 1 g
Formula Weight 222.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-iodopyridine