missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™

Catalog No. AC457590050
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457590050 5g
AC457590250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457590050 Supplier Thermo Scientific Chemicals Supplier No. 457590050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65145-13-3
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD01310985
InChI Key NXWTWYULZRDBSA-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid
PubChem CID 2783398
IUPAC Name 2-fluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-hydroxybenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

RUO – Research Use Only