missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH5517803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5517803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5517803 Supplier Thermo Scientific Chemicals Supplier No. H5517803
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula C12H10BFN2O3
MDL Number MFCD22682831

Specifications

Quantity 1 g
Formula Weight 260.03
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only