missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitropyridine, 98%

Catalog No. p-7053666
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6126014 25 g
AAH6126022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6126014 Supplier Thermo Scientific Chemicals Supplier No. H6126014
Only null left

CAS: 1480-87-1 | C5H3FN2O2 | 142.09 g/mol

2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1480-87-1
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.09
MDL Number MFCD03095068
InChI Key QDKIYDGHCFZBGC-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n
PubChem CID 2762817
IUPAC Name 2-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=C(F)N=CC=C1

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 110°C (10 mmHg)
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 2-Fluoro-3-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only