missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. F11051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11051G 1 g
F11055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11051G Supplier TCI America Supplier No. F11051G

Chemical Identifiers

CAS 452972-14-4
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.054
MDL Number MFCD00016604
InChI Key YYOZFGQOPNTVGM-UHFFFAOYSA-N
Synonym 2-fluoropyridine-3-boronic acid pinacol ester, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-3-boronic acid, pinacol ester, pyridine, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2-fluoropyridin-3-yl boronic acid pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-3-boronic pound inverted question markacid pound inverted question markpinacol pound inverted question markester, pubchem24045, 2-fluoropyridine-3-boronicacidpinacolester
PubChem CID 12067063
IUPAC Name 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)F

Specifications

Melting Point 47°C
Color White
Quantity 1 g
Formula Weight 223.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine