Learn More
2'-Fluoro-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
Description
2'-Fluoro-2'-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2'-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Fluoro-2′-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2′-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Solubility Information | Soluble in water. |
| IUPAC Name | (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol |
| Formula Weight | 269.23 |
| Percent Purity | 99% |
| Physical Form | Powder |
| Chemical Name or Material | 2'-Fluoro-2'-deoxyadenosine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only