missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-1-methylpyridinium p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America F022525G
Specifications
| 2-Fluoro-1-methylpyridinium p-Toluenesulfonate [Fluorinating Reagent] | |
| 132°C | |
| C13H14FNO3S | |
| 25 g | |
| HQWDKLAIDBOLFE-UHFFFAOYSA-M | |
| 2-fluoro-1-methylpyridin-1-ium;4-methylbenzenesulfonate | |
| 171631 | |
| ≥98.0% (T) |
| 58086-67-2 | |
| White | |
| MFCD00011983 | |
| 2-fluoro-1-methylpyridinium p-toluenesulfonate, 2-fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate, 2-fluoro-1-methylpyridinium, mukaiyama's reagent, 2-fluoro-1-methylpyridinium 4-toluenesulfonate, 2-fluoro-1-methylpyridinium p-toluenesulphonate, 2-fluoro-n-methylpyridinium toluene-4-sulfonate, 2-fluoro-1-methylpyridin-1-ium tosylate, 2-fluoro-n-methylpyridiniumtoluene-4-sulphonate, 2-fluoro-1-methyl-pyridin-1-ium; 4-methylbenzenesulfonate | |
| CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=CC=C1F | |
| 283.317 | |
| 283.32 | |
| Crystalline Powder |
Chemical Identifiers
| 58086-67-2 | |
| 283.317 | |
| HQWDKLAIDBOLFE-UHFFFAOYSA-M | |
| 171631 | |
| CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=CC=C1F |
| C13H14FNO3S | |
| MFCD00011983 | |
| 2-fluoro-1-methylpyridinium p-toluenesulfonate, 2-fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate, 2-fluoro-1-methylpyridinium, mukaiyama's reagent, 2-fluoro-1-methylpyridinium 4-toluenesulfonate, 2-fluoro-1-methylpyridinium p-toluenesulphonate, 2-fluoro-n-methylpyridinium toluene-4-sulfonate, 2-fluoro-1-methylpyridin-1-ium tosylate, 2-fluoro-n-methylpyridiniumtoluene-4-sulphonate, 2-fluoro-1-methyl-pyridin-1-ium; 4-methylbenzenesulfonate | |
| 2-fluoro-1-methylpyridin-1-ium;4-methylbenzenesulfonate |
Safety and Handling
TSCA : No