missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-1-methylpyridinium p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. F02255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02255G 5 g
F022525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02255G Supplier TCI America Supplier No. F02255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58086-67-2 | C13H14FNO3S | 283.317 g/mol

Chemical Identifiers

CAS 58086-67-2
Molecular Formula C13H14FNO3S
Molecular Weight (g/mol) 283.317
MDL Number MFCD00011983
InChI Key HQWDKLAIDBOLFE-UHFFFAOYSA-M
Synonym 2-fluoro-1-methylpyridinium p-toluenesulfonate, 2-fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate, 2-fluoro-1-methylpyridinium, mukaiyama's reagent, 2-fluoro-1-methylpyridinium 4-toluenesulfonate, 2-fluoro-1-methylpyridinium p-toluenesulphonate, 2-fluoro-n-methylpyridinium toluene-4-sulfonate, 2-fluoro-1-methylpyridin-1-ium tosylate, 2-fluoro-n-methylpyridiniumtoluene-4-sulphonate, 2-fluoro-1-methyl-pyridin-1-ium; 4-methylbenzenesulfonate
PubChem CID 171631
IUPAC Name 2-fluoro-1-methylpyridin-1-ium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=CC=C1F

Specifications

Melting Point 132°C
Color White
Quantity 5 g
Formula Weight 283.32
Percent Purity ≥98.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-1-methylpyridinium p-Toluenesulfonate [Fluorinating Reagent]