missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylthiophene 96.0+%, TCI America™

Catalog No. E09151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09151G 1 g
E09155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09151G Supplier TCI America Supplier No. E09151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4298-52-6
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.16
MDL Number MFCD03425129
InChI Key LWISLHRIEATKTM-UHFFFAOYSA-N
Synonym 2-ethynyl-thiophene, 2-thienylethine, 2-thiophenylacetylene, 2-thienyl acetylene, pubchem16123, thien-2-yl acetylene, thiophene, 2-ethynyl, thiophen-2-yl acetylene, 2-ethynylthiophene, acmc-209js9
PubChem CID 5246258
IUPAC Name 2-ethynylthiophene
SMILES C#CC1=CC=CS1

Specifications

Color Brown-Yellow
Boiling Point 46°C
Quantity 1 g
UN Number 1993
Formula Weight 108.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethynylthiophene
TSCA No
Recommended Storage Refrigerator