missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™
$150.98 - $447.38
Chemical Identifiers
| CAS | 347389-74-6 |
|---|---|
| Molecular Formula | C8H13BO2 |
| Molecular Weight (g/mol) | 152 |
| MDL Number | MFCD13182122 |
| InChI Key | ZEOXJIIUMORBNN-UHFFFAOYSA-N |
| Synonym | Ethynylboronic Acid Pinacol Ester |
| PubChem CID | 15891429 |
| IUPAC Name | 2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E1074200MG
|
TCI America
E1074200MG |
200 mg |
Each for $150.98
|
|
|||||
|
E10741G
|
TCI America
E10741G |
1 g |
Each for $447.38
|
|
|||||
Chemical Identifiers
| 347389-74-6 | |
| 152 | |
| ZEOXJIIUMORBNN-UHFFFAOYSA-N | |
| 15891429 | |
| B1(OC(C(O1)(C)C)(C)C)C#C |
| C8H13BO2 | |
| MFCD13182122 | |
| Ethynylboronic Acid Pinacol Ester | |
| 2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
| 347389-74-6 | |
| 140°C | |
| MFCD13182122 | |
| 3272 | |
| ZEOXJIIUMORBNN-UHFFFAOYSA-N | |
| 2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 15891429 | |
| ≥95.0% (GC) | |
| 2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| Yellow | |
| C8H13BO2 | |
| 200 mg | |
| Ethynylboronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C#C | |
| 152 | |
| 152.00 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator