missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyltoluene, 98+%
Description
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.
Solubility
Soluble in water (74.6 mg/L (25°C)).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.
Solubility
Soluble in water (74.6 mg/L (25°C)).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | -17°C |
| Density | 0.884 |
| Boiling Point | 164°C to 165°C |
| Flash Point | 53°C (127°F) |
| Refractive Index | 1.504 |
| Quantity | 5 g |
| UN Number | UN3295 |
| Beilstein | 1851237 |
| Solubility Information | Soluble in water (74.6mg/L (25°C)). |
| Formula Weight | 120.2 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304 |
| Hazard Statement | GHS H Statement H304-H226 May be fatal if swallowed and enters airways. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| EINECSNumber | 210-255-1 |
| RTECSNumber | XT2500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only