missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyltoluene, 98+%

Catalog No. AAL0202806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0202806 5 g
AAL0202814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0202806 Supplier Thermo Scientific Chemicals Supplier No. L0202806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 611-14-3 | C9H12 | 120.195 g/mol

2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

Solubility
Soluble in water (74.6 mg/L (25°C)).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 611-14-3
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009257
InChI Key HYFLWBNQFMXCPA-UHFFFAOYSA-N
Synonym 2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
PubChem CID 11903
ChEBI CHEBI:34276
IUPAC Name 1-ethyl-2-methylbenzene
SMILES CCC1=CC=CC=C1C

Specifications

Melting Point -17°C
Density 0.884
Boiling Point 164°C to 165°C
Flash Point 53°C (127°F)
Refractive Index 1.504
Quantity 5 g
UN Number UN3295
Beilstein 1851237
Solubility Information Soluble in water (74.6mg/L (25°C)).
Formula Weight 120.2
Percent Purity ≥98%
Chemical Name or Material 2-Ethyltoluene
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 210-255-1
RTECSNumber XT2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only