missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylthiophene, 99%

Catalog No. p-7021150
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337214 25 g
AAA1337222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1337214 Supplier Thermo Scientific Chemicals Supplier No. A1337214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 872-55-9 | C6H8S | 112.19 g/mol

It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 872-55-9
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00005461
InChI Key JCCCMAAJYSNBPR-UHFFFAOYSA-N
Synonym thiophene, 2-ethyl, 2-ethyl-thiophene, ethylthiophene, thiophene, ethyl, pubchem5536, acmc-209qjc, 2-ethylthiophene, 2-ethylthiophene, analytical standard, met 2-cl-4-metpyrimidine-6-carboxylate
PubChem CID 13388
IUPAC Name 2-ethylthiophene
SMILES CCC1=CC=CS1

Specifications

Density 0.991
Boiling Point 133°C to 134°C
Flash Point 21°C (69°F)
Odor Stench
Refractive Index 1.512
Quantity 25 g
UN Number UN1993
Beilstein 105402
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 112.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylthiophene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement

H225-H315-H319-H335

Highly flammable liquid and vapour.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-830-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only