missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethylpyrrole 96.0+%, TCI America™
$126.08 - $429.68
Chemical Identifiers
CAS | 1551-06-0 |
---|---|
Molecular Formula | C6H9N |
Molecular Weight (g/mol) | 95.15 |
MDL Number | MFCD00060477 |
InChI Key | XRPDDDRNQJNHLQ-UHFFFAOYSA-N |
PubChem CID | 137075 |
IUPAC Name | 2-ethyl-1H-pyrrole |
SMILES | CCC1=CC=CN1 |
Chemical Identifiers
1551-06-0 | |
95.15 | |
XRPDDDRNQJNHLQ-UHFFFAOYSA-N | |
2-ethyl-1H-pyrrole |
C6H9N | |
MFCD00060477 | |
137075 | |
CCC1=CC=CN1 |
Specifications
1551-06-0 | |
165°C | |
MFCD00060477 | |
2-ethylpyrrole, 1h-pyrrole, 2-ethyl, pyrrole,2-ethyl, 1h-pyrrole,2-ethyl, acmc-209dbz, #, 2-ethylpyrrole, analytical standard, 2-ethylpyrrole, technical grade | |
CCC1=CC=CN1 | |
95.15 | |
95.15 | |
Liquid |
Yellow | |
C6H9N | |
1 g | |
XRPDDDRNQJNHLQ-UHFFFAOYSA-N | |
2-ethyl-1H-pyrrole | |
137075 | |
≥96.0% (GC) | |
2-Ethylpyrrole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator