missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylpyrrole 96.0+%, TCI America™

Catalog No. E07135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07135G 5 g
E07131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07135G Supplier TCI America Supplier No. E07135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1551-06-0
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00060477
InChI Key XRPDDDRNQJNHLQ-UHFFFAOYSA-N
PubChem CID 137075
IUPAC Name 2-ethyl-1H-pyrrole
SMILES CCC1=CC=CN1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 g
Synonym 2-ethylpyrrole, 1h-pyrrole, 2-ethyl, pyrrole,2-ethyl, 1h-pyrrole,2-ethyl, acmc-209dbz, #, 2-ethylpyrrole, analytical standard, 2-ethylpyrrole, technical grade
Formula Weight 95.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylpyrrole
TSCA No
Recommended Storage Refrigerator