missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E08181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08181G Supplier TCI America Supplier No. E08181G
Only null left

Chemical Identifiers

CAS 90002-36-1
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD02093075
InChI Key QSSPYZOSTJDTTL-UHFFFAOYSA-N
PubChem CID 4100862
IUPAC Name (2-ethylphenyl)boronic acid
SMILES CCC1=CC=CC=C1B(O)O

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Synonym 2-ethylbenzeneboronic acid, 2-ethylphenyl boronic acid, 2-ethylphenyl boranediol, boronic acid, 2-ethylphenyl, 2-ethylphenylboronicacid, o-ethylphenylboronic acid, pubchem14050, 2-ethylphenylboranic acid, 2-ethylbenzene boronic acid
Formula Weight 149.98
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethylphenylboronic Acid (contains varying amounts of Anhydride)