missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylimidazole 98.0+%, TCI America™
$59.69 - $334.98
Chemical Identifiers
| CAS | 1072-62-4 |
|---|---|
| Molecular Formula | C5H8N2 |
| Molecular Weight (g/mol) | 96.13 |
| MDL Number | MFCD00005192 |
| InChI Key | PQAMFDRRWURCFQ-UHFFFAOYSA-N |
| Synonym | 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972 |
| PubChem CID | 66130 |
| IUPAC Name | 2-ethyl-1H-imidazole |
| SMILES | CCC1=NC=CN1 |
Chemical Identifiers
| 1072-62-4 | |
| 96.13 | |
| PQAMFDRRWURCFQ-UHFFFAOYSA-N | |
| 66130 | |
| CCC1=NC=CN1 |
| C5H8N2 | |
| MFCD00005192 | |
| 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972 | |
| 2-ethyl-1H-imidazole |
Specifications
| 1072-62-4 | |
| White-Yellow | |
| MFCD00005192 | |
| 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972 | |
| CCC1=NC=CN1 | |
| 96.13 | |
| 96.13 | |
| Crystalline Powder |
| 83°C | |
| C5H8N2 | |
| 25 g | |
| PQAMFDRRWURCFQ-UHFFFAOYSA-N | |
| 2-ethyl-1H-imidazole | |
| 66130 | |
| ≥98.0% (GC,T) | |
| 2-Ethylimidazole |
Safety and Handling
EINECSNumber : (5)-0382
TSCA : Yes