missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylimidazole, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 83.0°C to 87.0°C |
| Boiling Point | 268.0°C |
| Flash Point | 159°C |
| Assay Percent Range | 1% max. Imidazole (GC) |
| Packaging | Glass bottle |
| Quantity | 500 g |
| Beilstein | 23, 78 |
| Solubility Information | Solubility in water: 617g/L (20°C). Other solubilities: soluble in many organic solvents |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H StatementCauses serious eye damage.Causes skin irritation.Harmful if swallowed. |
| Precautionary Statement | GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.Immediately |
| EINECSNumber | 214-011-5 |
| TSCA | TSCA |
RUO – Research Use Only