missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylimidazole, 99%

Catalog No. AC219281000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219281000 100 g Glass bottle
AC219285000 500 g Glass bottle
AC219280025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC219281000 Supplier Thermo Scientific Chemicals Supplier No. 219281000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072-62-4 | C5H8N2 | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-ethyl-1H-imidazole
SMILES CCC1=NC=CN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Boiling Point 268.0°C
Flash Point 159°C
Assay Percent Range 1% max. Imidazole (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 78
Solubility Information Solubility in water: 617g/L (20°C). Other solubilities: soluble in many organic solvents
Formula Weight 96.13
Percent Purity 99%
Chemical Name or Material 2-Ethylimidazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 214-011-5
TSCA TSCA

RUO – Research Use Only