missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Vinyl Ether 97.0+%, TCI America™

Supplier:  TCI America I014810G

 View more versions of this product

Catalog No. I014810G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Ethylhexyl Vinyl Ether
Colorless
C10H20O
10 g
Isooctyl Vinyl Ether
CCCCC(CC)COC=C
156.269
156.27
Liquid
103-44-6
177°C
MFCD00053800
3271
DSSAWHFZNWVJEC-UHFFFAOYSA-N
3-(ethenoxymethyl)heptane
61004
≥97.0% (GC)

Chemical Identifiers

103-44-6
156.269
DSSAWHFZNWVJEC-UHFFFAOYSA-N
61004
CCCCC(CC)COC=C
C10H20O
MFCD00053800
Isooctyl Vinyl Ether
3-(ethenoxymethyl)heptane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0372&(2)-0575

RTECSNumber : KO0175000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions