missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Vinyl Ether 97.0+%, TCI America™

Catalog No. I014825G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I014825G 25 g
I014810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I014825G Supplier TCI America Supplier No. I014825G

Chemical Identifiers

CAS 103-44-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00053800
InChI Key DSSAWHFZNWVJEC-UHFFFAOYSA-N
Synonym Isooctyl Vinyl Ether
PubChem CID 61004
IUPAC Name 3-(ethenoxymethyl)heptane
SMILES CCCCC(CC)COC=C

Specifications

Color Colorless
Boiling Point 177°C
Quantity 25 g
UN Number 3271
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Vinyl Ether
EINECSNumber (2)-0372&(2)-0575
RTECSNumber KO0175000
TSCA No