missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl Thioglycolate 98.0+%, TCI America™
$55.84 - $165.56
Chemical Identifiers
| CAS | 7659-86-1 |
|---|---|
| Molecular Formula | C10H20O2S |
| Molecular Weight (g/mol) | 204.33 |
| MDL Number | MFCD00022081 |
| InChI Key | OWHSTLLOZWTNTQ-UHFFFAOYNA-N |
| Synonym | Thioglycolic Acid 2-Ethylhexyl Ester, Mercaptoacetic Acid 2-Ethylhexyl Ester, Octyl Thioglycolate, Thioglycolic Acid Octyl Ester, 2-Ethylhexyl Mercaptoacetate |
| PubChem CID | 24309 |
| IUPAC Name | 2-ethylhexyl 2-sulfanylacetate |
| SMILES | CCCCC(CC)COC(=O)CS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T061225ML
|
TCI America
T061225ML |
25 mL |
Each for $55.84
|
|
|||||
|
T0612500ML
|
TCI America
T0612500ML |
500 mL |
Each for $165.56
|
|
|||||
Chemical Identifiers
| 7659-86-1 | |
| 204.33 | |
| OWHSTLLOZWTNTQ-UHFFFAOYNA-N | |
| 24309 | |
| CCCCC(CC)COC(=O)CS |
| C10H20O2S | |
| MFCD00022081 | |
| Thioglycolic Acid 2-Ethylhexyl Ester, Mercaptoacetic Acid 2-Ethylhexyl Ester, Octyl Thioglycolate, Thioglycolic Acid Octyl Ester, 2-Ethylhexyl Mercaptoacetate | |
| 2-ethylhexyl 2-sulfanylacetate |
Specifications
| 7659-86-1 | |
| C10H20O2S | |
| 25 mL | |
| OWHSTLLOZWTNTQ-UHFFFAOYNA-N | |
| 2-ethylhexyl 2-sulfanylacetate | |
| 24309 | |
| ≥98.0% (GC) | |
| 2-Ethylhexyl Thioglycolate |
| Colorless | |
| MFCD00022081 | |
| Thioglycolic Acid 2-Ethylhexyl Ester, Mercaptoacetic Acid 2-Ethylhexyl Ester, Octyl Thioglycolate, Thioglycolic Acid Octyl Ester, 2-Ethylhexyl Mercaptoacetate | |
| CCCCC(CC)COC(=O)CS | |
| 204.33 | |
| 204.33 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1359
RTECSNumber : AI7255000
TSCA : Yes