missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl Thioglycolate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 500 mL |
| Formula Weight | 204.33 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Ethylhexyl Thioglycolate |
Safety and Handling
| EINECSNumber | (2)-1359 |
| RTECSNumber | AI7255000 |
| TSCA | Yes |