missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl Methacrylate (stabilized with MEHQ) 99.0+%, TCI America™
$33.56 - $55.92
Chemical Identifiers
| CAS | 688-84-6 |
|---|---|
| Molecular Formula | C12H22O2 |
| Molecular Weight (g/mol) | 198.306 |
| MDL Number | MFCD00009494 |
| InChI Key | WDQMWEYDKDCEHT-UHFFFAOYSA-N |
| Synonym | 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391 |
| PubChem CID | 12713 |
| IUPAC Name | 2-ethylhexyl 2-methylprop-2-enoate |
| SMILES | CCCCC(CC)COC(=O)C(=C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M059125ML
|
TCI America
M059125ML |
25 mL |
Each for $33.56
|
|
|||||
|
M0591500ML
|
TCI America
M0591500ML |
500 mL |
Each for $55.92
|
|
|||||
Chemical Identifiers
| 688-84-6 | |
| 198.306 | |
| WDQMWEYDKDCEHT-UHFFFAOYSA-N | |
| 12713 | |
| CCCCC(CC)COC(=O)C(=C)C |
| C12H22O2 | |
| MFCD00009494 | |
| 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391 | |
| 2-ethylhexyl 2-methylprop-2-enoate |
Specifications
| 688-84-6 | |
| 88°C | |
| MFCD00009494 | |
| 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391 | |
| CCCCC(CC)COC(=O)C(=C)C | |
| 198.306 | |
| 198.31 | |
| Liquid |
| Colorless | |
| C12H22O2 | |
| 25 mL | |
| WDQMWEYDKDCEHT-UHFFFAOYSA-N | |
| 2-ethylhexyl 2-methylprop-2-enoate | |
| 12713 | |
| ≥99.0% (GC) | |
| 2-Ethylhexyl Methacrylate (stabilized with MEHQ) |
Safety and Handling
EINECSNumber : (2)-1039
RTECSNumber : OZ4630000
TSCA : Yes