missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Glycidyl Ether 98.0+%, TCI America™

Catalog No. p-7128899
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E022925ML 25 mL
E0229500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E022925ML Supplier TCI America Supplier No. E022925ML

Chemical Identifiers

CAS 2461-15-6
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00005141
InChI Key BBBUAWSVILPJLL-UHFFFAOYSA-N
Synonym (2-Ethylhexyloxy)-2,3-epoxypropane
PubChem CID 17162
IUPAC Name 2-(2-ethylhexoxymethyl)oxirane
SMILES CCCCC(CC)COCC1CO1

Specifications

Color Yellow
Boiling Point 61°C
Quantity 25 mL
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Glycidyl Ether
EINECSNumber (2)-2426
RTECSNumber TZ3300000
TSCA Yes